Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethyldisiloxane, 98+%, ACROS Organics™

Click to view available options
Quantity:
10mL
100mL
500mL
2.5L
Packaging:
Glass bottle
107-46-0
C6H18OSi2
162.38
UQEAIHBTYFGYIE-UHFFFAOYSA-N
hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide
24764
CHEBI:78002
trimethyl(trimethylsilyloxy)silane
C[Si](C)(C)O[Si](C)(C)C
107-46-0
C6H18OSi2
162.38
UQEAIHBTYFGYIE-UHFFFAOYSA-N
hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide
24764
CHEBI:78002
trimethyl(trimethylsilyloxy)silane
C[Si](C)(C)O[Si](C)(C)C
Silylation reagent
Viscosity 0.5 mPa.s (25°C)
Flash Point −6°C
Specific Gravity 0.76
Boiling Point 101.0°C
Color Colorless
Refractive Index 1.3765 to 1.3785
Density 0.7600g/mL
Packaging Glass bottle
Melting Point -59.0°C
Infrared Spectrum Authentic
Quantity 100mL
Assay Percent Range 98% min. (GC)
Linear Formula (CH3)3SiOSi(CH3)3
Beilstein 04,III,1859
Fieser 08,240; 09,55; 10,196
Merck Index 15,4726
Solubility Information Solubility in water: insoluble.
Formula Weight 162.38
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material Hexamethyldisiloxane
Show More Show Less

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.

GHS Signal Word: Danger

EINECSNumber : 203-492-7