Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexamethyleneimine

Catalog No. H10401100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10401100ML 100 mL
H1040125ML 25 mL
H10401500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H10401100ML Supplier Sigma Aldrich Supplier No. H10401100ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Hexamethyleneimine (hmi) can be used in the preparation of:
  • Zeolitic catalysts such as mcm-22 and mcm-49
  • Sapo-35, a molecular sieve
  • Various microporous aluminosilicates and titanosilicates
  • Organic-inorganic hybrid pervoskiteit can also been used in the total synthesis of various natural products and bioactive molecules such as ()-balanol, indole estrogens, derivatives of daunorubicin and (+)-cetiedil

Specifications

CAS 111-49-9
Density 0.88 g/mL (at 25°C (literature))
Boiling Point 138°C (749 mmHg)
Molecular Formula C6H13N
Refractive Index n20/D 1.466 (literature)
Linear Formula C6H13N
MDL Number MFCD00006934
Quantity 100 mL
Synonym 1-Azacycloheptane; Azepane; HMI; Perhydroazepine; Hexahydro-1 H-azepine; Homopiperidine
Molecular Weight (g/mol) 99.17
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.