Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethyleneimine, 98+%

Catalog No. p-7027334
Click to view available options
Quantity:
100 mL
500 mL

CAS: 111-49-9 | C6H13N | 99.177 g/mol

Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-49-9
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.177
MDL Number MFCD00006934
InChI Key ZSIQJIWKELUFRJ-UHFFFAOYSA-N
Synonym hexamethyleneimine, homopiperidine, hexahydroazepine, hexahydro-1h-azepine, perhydroazepine, 1h-azepine, hexahydro, hexamethylenimine, azacycloheptane, azepan, hexamethylene imine
PubChem CID 8119
ChEBI CHEBI:32616
IUPAC Name azepane
SMILES C1CCCNCC1

Specifications

Melting Point -37°C
Density 0.88
Boiling Point 138°C to 139°C
Flash Point 22°C (71°F)
Odor Ammonia-like
Refractive Index 1.466
Quantity 100 mL
UN Number UN2493
Beilstein 1084
Solubility Information Fully miscible in water.
Formula Weight 99.18
Percent Purity ≥98%
Chemical Name or Material Hexamethyleneimine
Show More Show Less
Hazard Category H225-H300-H314-H331
Hazard Statement GHS H Statement
H225-H300-H314-H318-H312-H332
Highly flammable liquid and vapour.
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXAMETHYLENEIMINE
EINECSNumber 203-875-9
RTECSNumber CM3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.