Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexamethylenetetramine

Catalog No. 33233500GR Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
100 g
500 g
1 kg

Catalog No. 33-233-500GR

Supplier: Sigma Aldrich 33233500GR

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexamethylenetetramine

Specifications

CAS 100-97-0
Melting Point 280°C (subl.)
Molecular Formula C6H12N4
Linear Formula C6H12N4
MDL Number MFCD00006895
Quantity 500 g
Synonym 1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane; Hexamine; Methenamine; Urotropine
Molecular Weight (g/mol) 140.19 g/mol
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.