Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethylenetetramine, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18610799 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

Chemical Identifiers

CAS 100-97-0
Molecular Formula C6H12N4
Molecular Weight (g/mol) 140.19
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
IUPAC Name 1,3,5,7-tetraazatricyclo[3.3.1.1³,⁷]decane
SMILES C1N2CN3CN1CN(C2)C3

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.02
Packaging Poly Bottle
Quantity 2.5 kg
Percent Purity 0.99
Grade ACS

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.