Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethylphosphoric Triamide 98.0+%, TCI America™
SDP

Catalog No. H009525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H009525G 25 g
H0095100G 100 g
H0095500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. H009525G Supplier TCI America Supplier No. H009525G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 680-31-9
Molecular Formula C6H18N3OP
Molecular Weight (g/mol) 179.204
MDL Number MFCD00008303
InChI Key GNOIPBMMFNIUFM-UHFFFAOYSA-N
Synonym hexamethylphosphoramide, hmpa, hexametapol, hexamethylphosphoric triamide, hempa, hmpta, hexamethylphosphoric acid triamide, hexamethylphosphorictriamide, hexamethylphosphotriamide, phosphoric hexamethyltriamide
PubChem CID 12679
ChEBI CHEBI:24565
IUPAC Name N-[bis(dimethylamino)phosphoryl]-N-methylmethanamine
SMILES CN(C)P(=O)(N(C)C)N(C)C

Specifications

Color Yellow
Boiling Point 232°C
Quantity 25 g
Formula Weight 179.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Hexamethylphosphoric Triamide
EINECSNumber (2)-1930
RTECSNumber TD0875000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.