Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanal, 96%, stabilized

Catalog No. p-3736578
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle

CAS: 66-25-1 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-25-1, 59-02-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00007027
InChI Key JARKCYVAAOWBJS-UHFFFAOYSA-N
Synonym caproaldehyde, hexaldehyde, caproic aldehyde, capronaldehyde, 1-hexanal, n-hexanal, hexanaldehyde, n-caproaldehyde, hexylaldehyde, aldehyde c-6
PubChem CID 6184
IUPAC Name hexanal
SMILES CCCCCC=O

Specifications

Acidity 10mg KOH/g max.
Melting Point -56.0°C
Color Colorless to Yellow
Density 0.8150g/mL
Boiling Point 130.0°C to 131.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4020 to 1.4070
Linear Formula CH3(CH2)4CHO
Quantity 500 mL
Beilstein 01, II, 745
Specific Gravity 0.815
Merck Index 15, 1762
Solubility Information Solubility in water: 4.8g/L (20°C). Other solubilities: soluble in methanol, acetone, chloroform, ether
Stabilizer approx. 500ppm alpha-tocopherol
Viscosity 0.69 mPa.s (20°C)
Formula Weight 100.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Hexanal
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse caut
EINECSNumber 200-624-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.