Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanal, 98%

Catalog No. p-7023156
Change view
Click to view available options
Quantity:
100 g
500 g
Catalog No. Quantity
AAA1626522 100 g
AAA1626536 500 g
2 options

Catalog No. AAA1626522

Supplier: Thermo Scientific Chemicals A1626522

Only null left
Add to Cart

CAS: 66-25-1 | C6H12O | 100.161 g/mol

Hexanal is used in the flavor industry to prepare fruity flavors. It is utilized as a flavoring agent in the food industry. It is also used in Witting and aldol reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexanal is used in the flavor industry to prepare fruity flavors. It is utilized as a flavoring agent in the food industry. It is also used in Witting and aldol reactions.

Solubility
Miscible with ethanol, ethyl ether, acetone, benzene and water.

Notes
Air sensitive. Incompatible with oxidizing agents, strong acids, strong bases and strong reducing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-25-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00007027
InChI Key JARKCYVAAOWBJS-UHFFFAOYSA-N
Synonym caproaldehyde, hexaldehyde, caproic aldehyde, capronaldehyde, 1-hexanal, n-hexanal, hexanaldehyde, n-caproaldehyde, hexylaldehyde, aldehyde c-6
PubChem CID 6184
IUPAC Name hexanal
SMILES CCCCCC=O

Specifications

Melting Point °C to 56°C
Density 0.834 g/mL
Boiling Point 130°C to 131°C
Flash Point 25°C (77°F)
Assay Percent Range 98%
Refractive Index 1.405
Quantity 100 g
UN Number UN1207
Beilstein 506198
Sensitivity Air Sensitive
Merck Index 14,1760
Solubility Information Miscible with ethanol,ethyl ether,acetone,benzene and water.
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material Hexanal
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226-H315-H319-H335
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXALDEHYDE
EINECSNumber 200-624-5
RTECSNumber MN7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.