Learn More
Hexanal, 98%
Description
Hexanal is used in the flavor industry to prepare fruity flavors. It is utilized as a flavoring agent in the food industry. It is also used in Witting and aldol reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Hexanal is used in the flavor industry to prepare fruity flavors. It is utilized as a flavoring agent in the food industry. It is also used in Witting and aldol reactions.
Solubility
Miscible with ethanol, ethyl ether, acetone, benzene and water.
Notes
Air sensitive. Incompatible with oxidizing agents, strong acids, strong bases and strong reducing agents. Store in a cool place.
Specifications
Specifications
| Melting Point | °C to 56°C |
| Density | 0.834 g/mL |
| Boiling Point | 130°C to 131°C |
| Flash Point | 25°C (77°F) |
| Assay Percent Range | 98% |
| Refractive Index | 1.405 |
| Quantity | 100 g |
| UN Number | UN1207 |
| Beilstein | 506198 |
| Sensitivity | Air Sensitive |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319 |
| Hazard Statement | GHS H Statement H226-H315-H319-H335 |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXALDEHYDE |
| EINECSNumber | 200-624-5 |
| RTECSNumber | MN7175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.