Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.9% (>60% n-Hexane), Honeywell
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
19 L
4 x 4 L
204 L
Packaging:
Glass Bottle
Metal Can
Metal Drum
Poly Bottle
Specifications
Specifications
CAS Min % | >60.0000% |
CAS Max % | >60.00%, <40.0000% |
Density | 0.659 - 0.673 g/cm3 |
Boiling Point | 68.7°C |
Quantity | 204 L |
Flash Point | −26°C |
Packaging | Metal Drum |
UN Number | UN1208 |
Beilstein | 1730733 |
Water | 0.01% max. |
Show More |
Safety and Handling
ShelfLife | 547 days from date of manufacture |
DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction