Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexane, Laboratory Reagent, ≥95%, Honeywell™

Catalog No. 60045159
Click to view available options
Quantity:
1 L
2.5 L
20 L
4 x 2.5 L
4 x 5 L
6 x 1 L
190 L
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
Stainless Steel Can

Laboratory Reagent,≥95%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Color Colorless
Density 0.659g/mL at 25°C
Boiling Point 69°C
Quantity 190 L
Explosion Limit 0.077
Flash Point −26°C (−14.8°F)
Ignition Point 453°F
Packaging Metal Drum
Physical Form Liquid
Refractive Index n20/D 1.375(lit.)
Linear Formula CH3(CH2)4CH3
Vapor Pressure 132mmHg (20°C), 256mmHg (37.7°C), 5.2psi (37.7°C)
UN Number UN1208
Beilstein 1730733
Formula Weight 86.18g/mol
Percent Purity ≥95%
Grade Laboratory Reagent
Chemical Name or Material Hexane
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P370 + P378-P403 + P235
Precautionary Statement H225-H304-H315-H336-H361f-H373-H411
DOTInformation Class 3 - PG 2 - Hexanes
EINECSNumber 203-777-6
RTECSNumber MN9275000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.