Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Hexane-1 6-diammonium Iodide
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 g
Description
Organohalide based perovskites have emerged as an important class of material for solar cell applications. The variations/substitution in organohalide cations and anions is employed for the optimization of the band gap, carrier diffusion length, and power conversion efficiency of perovskites based solar cells.Product of Greatcell Solar(R)
Specifications
Specifications
| CAS | 20208-23-5 |
| Molecular Formula | C6H18I2N2 |
| Linear Formula | C6H18I2N2 |
| Quantity | 10 g |
| Synonym | 1,6-Hexanediamine, dihydriodide (8 CI,9 CI); Hexamethylenediamine dihydroiodide; Hexamethylenediammonium diiodide; Hexamethylenediammonium iodide; NHEXDAI |
| Molecular Weight (g/mol) | 372.03 |
| Biological Activity | Respiratory system |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction