Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexanes (Certified), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Glass Bottle
Steel Drum
Steel Pail
Specifications
Specifications
Melting Point | -95°C |
Color | Colorless |
Density | 0.659g/cm³ |
Boiling Point | 69°C |
Quantity | 4 L |
Flash Point | −6.7°C (20°F) |
Identification | Pass Test |
Packaging | Glass Bottle |
Physical Form | Liquid |
Residue after Evaporation | 0.001% max. |
Show More |
Safety and Handling


Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Extremely flammable liquid and vapor. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. NFPA Health:2 Flammability:3 Instability:0 |
DOTInformation | DOT Class 3, : Flammable Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction