Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanes, for HPLC, contains ca. 95% n-Hexane

Catalog No. AC268360025
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle

CAS: 92112-69-1 | C6H14

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hexanes
Name Note (95% n-Hexane)
Absorbance (1 cm cell vs HPLC−grade water), 0.005 max. at 250nm, 0.01 max. at 240nm, 0.03 max. at 230nm, 0.07 max. at 220nm, 0.25 max. at 210nm, 0.7 max. at 200nm
CAS 92112-69-1, 110-54-3
Density 0.6590g/mL
Quantity 2.5 L
Packaging Glass Bottle
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
Vapor Pressure Vapor Pressure: 160mbar at 20°C
MDL Number MFCD00009520
Beilstein 01, 142
Merck Index 14, 4694
Synonym Hex
Formula Weight 86.18
Grade HPLC
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.