Learn More
Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 210590050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexanes | |
Mixture of isomers | |
0.6590g/mL | |
5 L | |
Plastic Drum | |
1.3748 to 1.3810 | |
0.659 | |
CH3(CH2)4CH3 | |
0.31 mPa.s (20°C) | |
14, 4694 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform | |
Technical |
-95.0°C | |
92112-69-1, 110-54-3 | |
69.0°C | |
−22°C | |
Liquid | |
0.002% max. | |
C6H14 | |
MFCD00009520 | |
01, 142 | |
Hex | |
86.18 |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF exposed or concerned: Get medical advice/attention.
Danger
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only