Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanoic acid, 98+%

Catalog No. AAA13789AP
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 142-62-1 | C6H12O2 | 116.16 g/mol

Hexanoic acid is used to prepare esters by reacting with alcohols, which finds application in artificial flavors. It is also involved in the production of hexylphenols, hexanoates and caproates. Further, it is used as non-viral gene carrier as well as to protect tomato plants from Botrytis cinerea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexanoic acid is used to prepare esters by reacting with alcohols, which finds application in artificial flavors. It is also involved in the production of hexylphenols, hexanoates and caproates. Further, it is used as non-viral gene carrier as well as to protect tomato plants from Botrytis cinerea.

Solubility
Miscible with diethyl ether and ethanol. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents, reducing agents and allyl alcohol.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-62-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00004421
InChI Key FUZZWVXGSFPDMH-UHFFFAOYSA-N
Synonym caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid
PubChem CID 8892
ChEBI CHEBI:30776
IUPAC Name hexanoic acid
SMILES CCCCCC(=O)O

Specifications

Melting Point °C to 4°C
Density 0.929
Boiling Point 202°C to 203°C
Flash Point 104°C (219°F)
Assay Percent Range 99%
Refractive Index 1.4165
Linear Formula CH3(CH2)4CO2H
Quantity 500 mL
UN Number UN2829
Beilstein 773837
Merck Index 14,1759
Solubility Information Miscible with diethyl ether and ethanol. Slightly miscible with water.
Formula Weight 116.16
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material n-Hexanoic Acid
Show More Show Less
Hazard Category H302-H312-H314-H318-H332-H335
Hazard Statement GHS H Statement
H311-H331-H314-H318-H302
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CAPROIC ACID
EINECSNumber 205-550-7
RTECSNumber MO5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.