Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanoic anhydride, 98%

Catalog No. p-7027194
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 2051-49-2 | C12H22O3 | 214.305 g/mol

It is used as a reactant in the synthesis of acremomannolipin A. It is used in green synthesis of esters of acyclovir (acyclovir prodrugs), in the preparation of hexanoyl-modified chitosan nanoparticles, chitosan-based polymeric surfactants via N-acylation of chitosans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reactant in the synthesis of acremomannolipin A. It is used in green synthesis of esters of acyclovir (acyclovir prodrugs), in the preparation of hexanoyl-modified chitosan nanoparticles, chitosan-based polymeric surfactants via N-acylation of chitosans.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-49-2
Molecular Formula C12H22O3
Molecular Weight (g/mol) 214.305
MDL Number MFCD00009509
InChI Key PKHMTIRCAFTBDS-UHFFFAOYSA-N
Synonym hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride
PubChem CID 74918
IUPAC Name hexanoyl hexanoate
SMILES CCCCCC(=O)OC(=O)CCCCC

Specifications

Melting Point -40°C
Density 0.928
Boiling Point 246°C to 248°C
Flash Point 128°C (262°F)
Odor Unpleasant
Refractive Index 1.429
Linear Formula [CH3(CH2)4CO]2O
Quantity 50 g
UN Number UN3265
Beilstein 1776561
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 214.31
Percent Purity 98%
Chemical Name or Material Hexanoic anhydride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 218-121-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.