Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanoyl chloride, 97%

Catalog No. p-7026800
Click to view available options
Quantity:
100 g
500 g

CAS: 142-61-0 | C6H11ClO | 134.603 g/mol

Hexanoyl chloride has been used in the synthesis of (±)-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one, 14-methyl-1-octadecene. It is used as crystal liquid, pharmaceutical and organic synthesis material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexanoyl chloride has been used in the synthesis of (±)-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one, 14-methyl-1-octadecene. It is used as crystal liquid, pharmaceutical and organic synthesis material.

Solubility
Soluble in ether, chloroform.

Notes
Moisture sensitive. Store under inert gas. Incompatible with oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-61-0
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00000760
InChI Key YWGHUJQYGPDNKT-UHFFFAOYSA-N
Synonym caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid
PubChem CID 67340
IUPAC Name hexanoyl chloride
SMILES CCCCCC(=O)Cl

Specifications

Melting Point °C to 87°C
Density 0.963
Boiling Point 150°C to 153°C
Flash Point 50°C (122°F)
Assay Percent Range 0.98
Refractive Index 1.426
Quantity 100 g
UN Number UN2920
Sensitivity Moisture sensitive
Merck Index 14,1763
Solubility Information Soluble in ether,chloroform.
Formula Weight 134.61
Percent Purity 98%
Chemical Name or Material Hexanoyl chloride
Show More Show Less
Hazard Category H226-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226-H302-H332
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 205-549-1
TSCA Yes
storageNote1 Moisture Sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.