Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexanoyl-Lysine adduct Antibody (5D9), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259362PECY55
Description
Hexanoyl-Lysine adduct Monoclonal specifically detects Hexanoyl-Lysine adduct in Non-species specific samples. It is validated for Flow Cytometry.Specifications
Hexanoyl-Lysine adduct | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
Synthetic Hexanoyl modified Keyhole Limpet Hemocyanin (KLH). | |
0.1 mL | |
Specific for Hexanoyl-Lysine adduct (HEL) modified peptides and proteins. Does not detect free Hexanoyl-Lysine. Does not cross-react with Acrolein, Crotonaldehyde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
5D9 | |
Flow Cytometry | |
Mouse | |
Protein G purified | |
Primary | |
Non-species specific | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction