Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanoyl-Lysine adduct Mouse, HRP, Clone:5D9, Abnova™

Catalog No. 89334261 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-261 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-261 Supplier Abnova Corporation Supplier No. MAB17393
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic Hexanoyl-Lysine adduct (HEL).

Specifications

Antigen Hexanoyl-Lysine adduct
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5D9
Conjugate HRP
Dilution ELISA (1:1000)
Flow Cytometry (1:50)
Immunocytochemistry (1:50)
Immunofluorescence (1:50)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Host Species Mouse
Immunogen Synthetic Hexanoyl modified Keyhole Limpet Hemocyanin (KLH).
Quantity 100 μg
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.