Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexaphenyldisilane, 98%

Catalog No. AAL0492803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0492803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0492803 Supplier Thermo Scientific Chemicals Supplier No. L0492803

CAS: 1450-23-3 | C36H30Si2 | 518.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1450-23-3
Molecular Formula C36H30Si2
Molecular Weight (g/mol) 518.81
MDL Number MFCD00039558
InChI Key ZMHATUZXFSOVSC-UHFFFAOYSA-N
PubChem CID 74059
SMILES C1=CC=C(C=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 368°C to 370°C
Quantity 1 g
Beilstein 2024230
Synonym hexaphenyldisilane, disilane, hexaphenyl, 1,1,1,2,2,2-hexaphenyldisilane, disilane, 1,1,1,2,2,2-hexaphenyl, disilane, hexaphenyl-,, acmc-1bxza, triphenyl triphenylsilyl silane, 1,1,1,2,2,2-hexaphenyldisilane #, disilane,1,1,1,2,2,2-hexaphenyl
IUPAC Name triphenyl(triphenylsilyl)silane
Formula Weight 518.81
Percent Purity 98%
Chemical Name or Material Hexaphenyldisilane
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 215-913-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.