Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HexArmor™ NXT™ 302 Series Gloves

Catalog No. 19163508 Shop All Performance Fabrics, Inc Products
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Food service safety gloves offer ANSI/ISEA Cut Level A7 protection to help keep employees safe without sacrificing dexterity or comfort.

  • 3-finger cut resistant SuperFabric™ brand material coverage targets primary knife strike areas
  • High dexterity Coretek™ high performance shell keeps hands cool and comfortable during work
  • Meets European food migration testing (EN 1186:2002 SI 1998 No 1376)
  • Designed to be worn under standard nitrile, latex or vinyl gloves
  • Ambidextrous, sold in each quantity
  • Launderable

Specifications

Certifications/Compliance (Europe) EN 1186:2002 SI 1998 No 1376
Coating Material Nitrile
Color Gray, White
Cut Resistance Rating ANSI A7
Hand Ambidextrous
Material SuperFabric
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN 388:4X42A
For Use With (Application) Food service, cutting and processing, deboning applications, knife/blade handling, Plastics trimming
Glove Cuff Style Knit
Model 10-302/S
Series NXT™ 10-302
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.