Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexavalent Chromium, standard solution, Specpure™ Cr∣+6 1000μg/mL
Click to view available options
Quantity:
100 mL
500 mL
Description
Hexavalent Chromium, standard solution is used in ion chromatography for measurement and determination of soluble Cr(VI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Odor | Odorless |
| Physical Form | Liquid |
| Color | Yellow |
| Solubility | Miscible with water |
| Quantity | 100 mL |
| Grade | Specpure™ |
| Chemical Name or Material | Hexavalent Chromium standard solution, Cr+6 1000μg/mL |
Safety and Handling
| Hazard Category | H340-H350-H360 |
| Hazard Statement | GHS H Statement H334-H340-H350-H360-H317-H208 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 232-143-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction