Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HEXB Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300084PE
Description
HEXB Polyclonal antibody specifically detects HEXB in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
HEXB | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
beta-hexosaminidase subunit beta, Beta-N-acetylhexosaminidase subunit beta, Cervical cancer proto-oncogene 7 protein, EC 3.2.1.52, ENC-1AS, HCC-7, hexosaminidase B (beta polypeptide), Hexosaminidase subunit B, N-acetyl-beta-glucosaminidase subunit beta | |
Produced in rabbits immunized with purified, recombinant Human HEXB (Accession#: P07686; Ala43-Met556) | |
0.1 mL | |
Neurodegeneration | |
3074 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction