Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEXIM1 Mouse anti-Human, Clone: 2C8A2, Proteintech
SDP

Catalog No. 501736743 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6743 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6743 Supplier Proteintech Group Inc Supplier No. 663111IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

66311-1-Ig recognizes the 54 kDa band.

The protein encoded by this gene is a member of the RecQ DNA helicase family. DNA helicases are enzymes involved in various types of DNA repair, including mismatch repair, nucleotide excision repair and direct repair. Some members of this family are associated with genetic disorders with predisposition to maligcy and chromosomal instability. The biological function of this helicase has not yet been determined. Two alternatively spliced transcripts, which encode the same isoform but differ in their 5′ and 3′ UTRs, have been described.

Specifications

Antigen HEXIM1
Applications Western Blot
Classification Monoclonal
Clone 2C8A2
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HEXIM1
Gene Accession No. O94992
Gene Alias Cardiac lineage protein 1, CLP1, EDG1, HEXIM1, HIS1, MAQ1, Menage a quatre protein 1, Protein HEXIM1
Gene Symbols HEXIM1
Host Species Mouse
Immunogen HEXIM1 Fusion Protein Ag8019
Purification Method Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10614
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.