Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEXIM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552629
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552629 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552629 Supplier Invitrogen™ Supplier No. PA552629
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKSDDTSDDD FMEEGGEEDG GSDGMGGDGS EFLQRDFSET YERYHTESLQ NMSKQELIKE YLELEKCLSR MEDENNRLRL ESKRLGGDDA RVRELELELD RLRAENLQLL TENELHRQQE RA Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

The protein encoded by this gene is a member of the RecQ DNA helicase family. DNA helicases are enzymes involved in various types of DNA repair, including mismatch repair, nucleotide excision repair and direct repair. Some members of this family are associated with genetic disorders with predisposition to malignancy and chromosomal instability. The biological function of this helicase has not yet been determined. Two alternatively spliced transcripts, which encode the same isoform but differ in their 5' and 3' UTRs, have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEXIM1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HEXIM1
Gene Accession No. O94992
Gene Alias 7330426E13Rik; Cardiac lineage protein 1; CLP1; Clp-1; EDG1; Estrogen down-regulated gene 1 protein; FLJ13562; hexamethylene bisacetamide inducible 1; hexamethylene bis-acetamide inducible 1; hexamethylene bisacetamide-inducible protein; hexamethylene bis-acetamide-inducible protein 1; hexamethylene-bis-acetamide-inducible transcript 1; hexamthylene bis-acetamide inducible 1; HEXIM1; HGNC:24953; HIS1; HMBA-inducible; MAQ1; menage a quatre 1; menage a quatre protein 1; Protein HEXIM1
Gene Symbols HEXIM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HEXIM1. Recombinant protein control fragment (Product #RP-89427).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10614
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.