Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HEXIM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538554
Description
Expression of this gene is induced by hexamethylene-bis-acetamide in vascular smooth muscle cells. This gene has no introns.
Specifications
| HEXIM1 | |
| Polyclonal | |
| Unconjugated | |
| HEXIM1 | |
| HEXIM1, hexamethylene bis-acetamide inducible 1, HGNC:24953, CLP1, HIS1, MAQ1 , HMBA-inducible, hexamethylene bisacetamide-inducible protein, hexamethylene-bis-acetamide-inducible transcript 1, hexamthylene bis-acetamide inducible 1, menage a quatre 1, EDG1, FLJ13562 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10614, 192231, 498008 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O94992, Q8R409, Q5M9G1 | |
| HEXIM1 | |
| A synthetic peptide derived from the internal region of human HES7 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction