Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEXIM1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989085
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9085 Supplier Bioss Supplier No. BS9460R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of the RecQ DNA helicase family. DNA helicases are enzymes involved in various types of DNA repair, including mismatch repair, nucleotide excision repair and direct repair. Some members of this family are associated with genetic disorders with predisposition to malignancy and chromosomal instability. The biological function of this helicase has not yet been determined. Two alternatively spliced transcripts, which encode the same isoform but differ in their 5′ and 3′ UTRs, have been described.

Specifications

Antigen HEXIM1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HEXIM1
Gene Accession No. O94992
Gene Alias 7330426E13Rik; Cardiac lineage protein 1; Clp1; CLP-1; EDG1; Estrogen down-regulated gene 1 protein; FLJ13562; hexamethylene bisacetamide inducible 1; hexamethylene bis-acetamide inducible 1; hexamethylene bisacetamide-inducible protein; Hexamethylene bis-acetamide-inducible protein 1; hexamethylene-bis-acetamide-inducible transcript 1; hexamthylene bis-acetamide inducible 1; Hexim1; HGNC:24953; HIS1; HMBA-inducible; MAQ1; menage a quatre 1; Menage a quatre protein 1; Protein HEXIM1
Gene Symbols HEXIM1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HEXIM1/EDG1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10614
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.