Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEXIM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554764
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554764 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554764 Supplier Invitrogen™ Supplier No. PA554764
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELVRDYLELE KRLSQAEEET RRLQQLQACT GQQSCRQVEE LAAEVQRLRT ENQRLRQENQ MWNREGCRCD EEPGT Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.

Transcriptional regulator which functions as a general RNA polymerase II transcription inhibitor. In cooperation with 7SK snRNA sequesters P-TEFb in a large inactive 7SK snRNP complex preventing RNA polymerase II phosphorylation and subsequent transcriptional elongation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEXIM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HEXIM2
Gene Accession No. Q96MH2
Gene Alias 4933402L21Rik; hexamethylene bisacetamide inducible 2; hexamethylene bis-acetamide inducible 2; hexamethylene bis-acetamide-inducible protein 2; hexamethylene-bis-acetamide-inducible transcript 2; hexamthylene bis-acetamide inducible 2; HEXIM2; L3; MAQ1 paralog; protein HEXIM2; RGD1311635
Gene Symbols HEXIM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HEXIM2. Recombinant protein control fragment (Product #RP-93398).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124790
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.