Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HEXO Antibody (OTI1B3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP201655
Description
HEXO Monoclonal specifically detects HEXO in Human, Rat, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HEXO | |
Monoclonal | |
1.1 mg/mL | |
Western Blot 1:500-2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
ERI1 | |
Full length human recombinant protein of human ERI1(NP_699163) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
90459 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI1B3 | |
Unconjugated | |
3' exoribonuclease, 3'-5' exonuclease ERI1, 3'EXO, 3'HEXO, EC 3.1, enhanced RNAi three prime mRNA exonuclease homolog 1, Eri-1 homolog, exoribonuclease 1, HEXO, histone mRNA 3' end-specific exonuclease, Histone mRNA 3'-end-specific exoribonuclease, Histone mRNA 3'-exonuclease 1, MGC35395, Protein 3'hExo, THEX1, three prime histone mRNA exonuclease 1,3'-5' exoribonuclease 1, three prime mRNA exonuclease 1 | |
Mouse | |
39.9 kDa | |
0.1 mL | |
Primary | |
Human, Rat, Primate, Monkey | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction