Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexokinase 1 Antibody (4D7), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104295IR
Description
Hexokinase 1 Monoclonal specifically detects Hexokinase 1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Hexokinase 1 | |
Monoclonal | |
DyLight 755 | |
P19367 | |
HK1 | |
Recombinant human Hexokinase1 (1-917aa) purified from E. coli | |
0.1 mL | |
Cancer | |
3098 | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
4D7 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Brain form hexokinase, EC 2.7.1, EC 2.7.1.1, glycolytic enzyme, hexokinase 1, Hexokinase type I, hexokinase-1, HK I, HK1-ta, HK1-tb, HK1-tc, HKI, HXK1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
The antibody recognizes all four isoforms of Hexokinase (1∼4) in recombinant protein. | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction