Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexokinase 2 Antibody (1A7), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104296IR
Description
Hexokinase 2 Monoclonal specifically detects Hexokinase 2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Hexokinase 2 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HK2 | |
Recombinant human Hexokinase 2 (1-917aa) purified from E. coli | |
0.1 mL | |
Cell Cycle and Replication | |
3099 | |
Human | |
Purified |
Western Blot, ELISA | |
1A7 | |
Western Blot, ELISA | |
DKFZp686M1669, EC 2.7.1, hexokinase 2, Hexokinase type II, HKII, HXK2, muscle, Muscle form hexokinase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human Hexokinase 2 mAb, clone 1A7, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Hexokinase 2 protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction