Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexokinase 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP244234
Description
Specifically detects Hexokinase 2 in Yeast samples, and it is validated for ELISA, Immunohistochemistry, Western Blotting
Hexokinase 2 Polyclonal specifically detects Hexokinase 2 in Yeast samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
| Hexokinase 2 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| DKFZp686M1669, EC 2.7.1, hexokinase 2, Hexokinase type II, HKII, HXK2, muscle, Muscle form hexokinase | |
| Rabbit | |
| 53.74 kDa | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 3099 | |
| Yeast | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1:1000-1:5000, ELISA 1:3000-1:10000, Immunohistochemistry 1:10-1:500 | |
| P04806 | |
| HK2 | |
| Hexokinase 2 Antibody was produced by repeated immunizations with yeast hexokinase protein. (Uniprot: P04806) | |
| Multi-step | |
| RUO | |
| Primary | |
| Reconstitute with 100 ul deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction