Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexokinase 2 Antibody (OTI4C5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP202272
Description
Hexokinase 2 Monoclonal specifically detects Hexokinase 2 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Hexokinase 2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
HK2 | |
Full length human recombinant protein of human HK2 (NP_000180) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
Human Hexokinase-2 protein is 74% identical with Hexokinase-1 and our Hexokinase 2 antibody (clone 4C5) is not expected to crossreact with Hexokinase-1. | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI4C5 | |
Unconjugated | |
DKFZp686M1669, EC 2.7.1, hexokinase 2, Hexokinase type II, HKII, HXK2, muscle, Muscle form hexokinase | |
Mouse | |
102.4 kDa | |
0.1 mL | |
Cell Cycle and Replication | |
3099 | |
Human, Mouse, Rat, Canine | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction