Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl acetate, 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
1L
2.5L
Packaging:
Glass bottle
142-92-7
C8H16O2
144.21
AOGQPLXWSUTHQB-UHFFFAOYSA-N
n-hexyl acetate,1-hexyl acetate,hexyl ethanoate,acetic acid hexyl ester,n-hexyl ethanoate,acetic acid, hexyl ester,hexyl alcohol, acetate,l-hexyl acetate,fema no. 2565,natural
8908
CHEBI:87510
hexyl acetate
CCCCCCOC(=O)C
142-92-7
C8H16O2
144.21
AOGQPLXWSUTHQB-UHFFFAOYSA-N
n-hexyl acetate,1-hexyl acetate,hexyl ethanoate,acetic acid hexyl ester,n-hexyl ethanoate,acetic acid, hexyl ester,hexyl alcohol, acetate,l-hexyl acetate,fema no. 2565,natural
8908
CHEBI:87510
hexyl acetate
CCCCCCOC(=O)C
Infrared Spectrum Authentic
Acidity 1mg KOH/g max.
Density 0.8710g/mL
Packaging Glass bottle
Color Colorless to Yellow
Quantity 1L
Specific Gravity 0.871
Refractive Index 1.4070 to 1.4110
Melting Point -80.0°C
Boiling Point 168.0°C to 170.0°C
Flash Point 43°C
Assay Percent Range 99%
Linear Formula CH3CO2C6H13
Beilstein 02,132
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in alcohol and ether
Formula Weight 144.21
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Hexyl acetate
Show More Show Less