Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl Chloroformate 98.0+%, TCI America™
SDP

Catalog No. C072325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C072325ML 25 mL
C07235ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C072325ML Supplier TCI America Supplier No. C072325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6092-54-2
Molecular Formula C7H13ClO2
Molecular Weight (g/mol) 164.63
MDL Number MFCD00000651
InChI Key KIWBRXCOTCXSSZ-UHFFFAOYSA-N
Synonym hexyl chloroformate, carbonochloridic acid, hexyl ester, n-hexyl chloroformate, chloroformic acid n-hexyl ester, hexyl chlorocarbonate, unii-mu6004c2z3, chloro hexyloxy methanone, chloroformic acid hexyl ester, hexylchloroformate, hexyl=chloroformate
PubChem CID 22466
IUPAC Name hexyl carbonochloridate
SMILES CCCCCCOC(Cl)=O

Specifications

Color Colorless
Boiling Point 78°C
Quantity 25 mL
UN Number 3277
Formula Weight 164.63
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Hexyl Chloroformate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.