Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexylene Glycol, Reagent, 98%, Spectrum™ Chemical
SDP

Catalog No. 18603131 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-131 20 L Amber Glass Bottle
18-603-129 500 mL Amber Glass Bottle
18-603-130 4 L Amber Glass Bottle
18-603-132 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-131 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HE18820LT
Add to Cart
Add to Cart

HE188, 107-41-5, C6H14O2

Spectrum™ Chemical Hexylene Glycol, Reagent

Chemical Identifiers

CAS 107-41-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
InChI Key SVTBMSDMJJWYQN-UHFFFAOYNA-N
IUPAC Name 2-methylpentane-2,4-diol
SMILES CC(O)CC(C)(C)O

Specifications

CAS Max % 100%
Boiling Point 197°C (386.6°F)
Packaging Amber Glass Bottle
Quantity 20 L
Percent Purity 98%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Hexylene Glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.