Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEY1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$160.07 - $480.74

Specifications

Antigen HEY1
Concentration 0.5 mg/mL
Dilution Western Blot 2-4ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227152SS
SDP
View Documents
Novus Biologicals
NBP227152SS
0.025 mg
Each for $160.07
Only null left
Add to Cart
 
NBP227152
SDP
View Documents
Novus Biologicals
NBP227152
0.1 mg
Each for $480.74
Only null left
Add to Cart
 
Description

Description

HEY1 Polyclonal specifically detects HEY1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

HEY1
Western Blot 2-4ug/ml
Polyclonal
Purified
RUO
Mouse
Q9Y5J3
23462
A portion of amino acids 50-100 of mouse Hey1 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
DNA Repair, DNA replication Transcription Translation and Splicing, Neuroscience, Transcription Factors and Regulators
PBS and 0.05% BSA with 0.05% Sodium Azide
basic helix-loop-helix protein OAF1, BHLHb31, Cardiovascular helix-loop-helix factor 2, CHF-2, CHF2hHRT1, Class B basic helix-loop-helix protein 31, Hairy and enhancer of split-related protein 1, hairy/enhancer-of-split related with YRPW motif 1, hairy/enhancer-of-split related with YRPW motif protein 1, Hairy-related transcription factor 1, HERP2HES-related repressor protein 1, HESR-1, HESR1MGC1274, HES-related repressor protein 2, HRT1, HRT-1OAF1
HEY1
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, mouse and rat.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.