Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HEY1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531076
Description
Recommended positive controls: DDDDK-tagged HEY1-transfected 293T. Predicted reactivity: Mouse (96%), Rat (89%), Xenopus laevis (81%), Dog (94%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Hairy/enhancer-of-split related with YRPW motif protein 1 belongs to the HEY family and is a Notch Signaling Pathway Protein. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. It is a downstream effector of Notch signaling which may be required for cardiovascular development. It acts as transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. It represses transcription by the cardiac transcriptional activators GATA4 and GATA6. Hey1 is also reported to be expressed during development of the nervous system.
Specifications
| HEY1 | |
| Polyclonal | |
| Unconjugated | |
| HEY1 | |
| AI316788; AI414254; basic helix-loop-helix protein OAF1; basic helix-loop-helix transcription factor; BC8; bHLH protein Hesr-1/Hey1; bHLHb31; cardiovascular basic helix-loop-helix factor 2; cardiovascular helix-loop-helix factor 2; CHF2; CHF-2; class B basic helix-loop-helix protein 31; Hairy and enhancer of split-related protein 1; Hairy/E(spl)-related with YRPW motif 1; hairy/enhancer-of-split related with YRPW motif 1; hairy/enhancer-of-split related with YRPW motif protein 1; hairy-related transcription factor 1; Herp1; Herp2; hes related family bHLH transcription factor with YRPW motif 1; Hesr1; HESR-1; hes-related family bHLH transcription factor with YRPW motif 1; HES-related repressor protein 1; HES-related repressor protein 2; HEY1; hey1 protein; hHRT1; HRT1; HRT-1; id:ibd1292; MGC1274; mHRT1; OAF1; zgc:110572 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23462 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.04 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9Y5J3 | |
| HEY1 | |
| Recombinant protein encompassing a sequence within the center region of human HEY1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction