Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEY1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568563
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568563 Supplier Invitrogen™ Supplier No. PA568563
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Dog: 79%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 100%.

Hairy/enhancer-of-split related with YRPW motif protein 1 belongs to the HEY family and is a Notch Signaling Pathway Protein. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. It is a downstream effector of Notch signaling which may be required for cardiovascular development. It acts as transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. It represses transcription by the cardiac transcriptional activators GATA4 and GATA6. Hey1 is also reported to be expressed during development of the nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEY1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HEY1
Gene Accession No. 0
Gene Alias AI316788; AI414254; basic helix-loop-helix protein OAF1; basic helix-loop-helix transcription factor; BC8; bHLH protein Hesr-1/Hey1; bHLHb31; cardiovascular basic helix-loop-helix factor 2; cardiovascular helix-loop-helix factor 2; CHF2; CHF-2; class B basic helix-loop-helix protein 31; Hairy and enhancer of split-related protein 1; Hairy/E(spl)-related with YRPW motif 1; hairy/enhancer-of-split related with YRPW motif 1; hairy/enhancer-of-split related with YRPW motif protein 1; hairy-related transcription factor 1; Herp1; Herp2; hes related family bHLH transcription factor with YRPW motif 1; Hesr1; HESR-1; hes-related family bHLH transcription factor with YRPW motif 1; HES-related repressor protein 1; HES-related repressor protein 2; HEY1; hey1 protein; hHRT1; HRT1; HRT-1; id:ibd1292; MGC1274; mHRT1; OAF1; zgc:110572
Gene Symbols HEY1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human HEY1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155437
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.