Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEY1 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992146
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2146 25 μL
50-199-2145 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2146 Supplier Rockland Immunochemicals Supplier No. 600401HB5S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human HEY1. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to mouse and 91.7% with dog and cattle.

Hairy/enhancer-of-split related with YRPW motif protein 1 belongs to the HEY family and is a Notch Signaling Pathway Protein. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. It is a downstream effector of Notch signaling which may be required for cardiovascular development. It acts as transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. It represses transcription by the cardiac transcriptional activators GATA4 and GATA6. Hey1 is also reported to be expressed during development of the nervous system.

Specifications

Antigen HEY1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.06 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene HEY1
Gene Accession No. Q9WV93, Q9Y5J3
Gene Alias AI316788; AI414254; basic helix-loop-helix protein OAF1; basic helix-loop-helix transcription factor; BC8; bHLH protein Hesr-1/Hey1; BHLHB31; BOS_13914; cardiovascular basic helix-loop-helix factor 2; Cardiovascular helix-loop-helix factor 2; CHF2; CHF-2; Class B basic helix-loop-helix protein 31; Hairy and enhancer of split-related protein 1; Hairy/E(spl)-related with YRPW motif 1; hairy/enhancer-of-split related with YRPW motif 1; Hairy/enhancer-of-split related with YRPW motif protein 1; Hairy-related transcription factor 1; Herp1; HERP2; hes related family bHLH transcription factor with YRPW motif 1; HESR1; HESR-1; hes-related family bHLH transcription factor with YRPW motif 1; HES-related repressor protein 1; HES-related repressor protein 2; hey1; hey1 protein; hHRT1; HRT1; HRT-1; id:ibd1292; LOC100344717; MGC1274; mHRT1; OAF1; zgc:110572
Gene Symbols HEY1
Host Species Rabbit
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15213, 23462
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.