Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HEY1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310943100UL
Description
HEY1 Polyclonal specifically detects HEY1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| HEY1 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| basic helix-loop-helix protein OAF1, BHLHb31, Cardiovascular helix-loop-helix factor 2, CHF-2, CHF2hHRT1, Class B basic helix-loop-helix protein 31, Hairy and enhancer of split-related protein 1, hairy/enhancer-of-split related with YRPW motif 1, hairy/enhancer-of-split related with YRPW motif protein 1, Hairy-related transcription factor 1, HERP2HES-related repressor protein 1, HESR-1, HESR1MGC1274, HES-related repressor protein 2, HRT1, HRT-1OAF1 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human HEY1 (NP_036390). Peptide sequence PQNGHGNAGTTASPTEPHHQGRLGSAHPEAPALRAPPSGSFGPVLPVVTS | |
| 100 μg | |
| DNA Repair, DNA replication Transcription Translation and Splicing, Neuroscience, Transcription Factors and Regulators | |
| 23462 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction