Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567365
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567365 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567365 Supplier Invitrogen™ Supplier No. PA567365
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PCEETTSESDM DETIDVGSEN NYSGQSTSSV IRLNSPTTTS QIMARKK Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.

Downstream effector of Notch signaling which may be required for cardiovascular development. Transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. Represses transcription by the cardiac transcriptional activators GATA4 and GATA6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEY2
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Hey2
Gene Accession No. Q9UBP5
Gene Alias basic helix-loop-helix transcription factor; bHLHb32; cardiovascular basic helix-loop-helix factor 1; cardiovascular helix-loop-helix factor 1; Chf1; Class B basic helix-loop-helix protein 32; gridlock; GRL; h hHRT2; hairy and enhancer of split related 2; hairy and enhancer of split-related protein 2; hairy/enhancer-of-split related with YRPW motif 2; hairy/enhancer-of-split related with YRPW motif protein 2; Hairy-E(spl)-related with YRPW motif 2; hairy-related transcription factor 2; hCHF1; HERP; Herp1; Herp2; hes related family bHLH transcription factor with YRPW motif 2; hesr2; HESR-2; hes-related family bHLH transcription factor with YRPW motif 2; HES-related repressor protein 1; HES-related repressor protein 2; hey2; hHRT2; Hrt2; HRT-2; mHRT2; protein gridlock; Protein gridlock homolog; zgc:136746
Gene Symbols Hey2
Host Species Rabbit
Immunogen Recombinant Human HEY2. Recombinant protein control fragment (Product #RP-108022).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23493
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.