Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572676
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572676 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572676 Supplier Invitrogen™ Supplier No. PA572676
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HEY2 Polyclonal Antibody for Western Blot

Downstream effector of Notch signaling which may be required for cardiovascular development. Transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. Represses transcription by the cardiac transcriptional activators GATA4 and GATA6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEY2
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Hey2
Gene Accession No. Q9I9L0
Gene Alias basic helix-loop-helix transcription factor; bHLHb32; cardiovascular basic helix-loop-helix factor 1; cardiovascular helix-loop-helix factor 1; Chf1; Class B basic helix-loop-helix protein 32; gridlock; GRL; h hHRT2; hairy and enhancer of split related 2; hairy and enhancer of split-related protein 2; hairy/enhancer-of-split related with YRPW motif 2; hairy/enhancer-of-split related with YRPW motif protein 2; Hairy-E(spl)-related with YRPW motif 2; hairy-related transcription factor 2; hCHF1; HERP; Herp1; Herp2; hes related family bHLH transcription factor with YRPW motif 2; hesr2; HESR-2; hes-related family bHLH transcription factor with YRPW motif 2; HES-related repressor protein 1; HES-related repressor protein 2; hey2; hHRT2; Hrt2; HRT-2; mHRT2; protein gridlock; Protein gridlock homolog; zgc:136746
Gene Symbols Hey2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 224-260 amino acids from the C-terminal region of Zebrafish hey2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58146
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.