Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEYL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110632
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110632 Supplier Invitrogen™ Supplier No. PA5110632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLEGPSSRAD PVRIRLLSHL NSYAAEMEPS PTPTGPLAFP AWPWSFFHSC PGLPALSNQL AILGRVPSPV LPGVSSPAYP IPALRTAPLR RATGIILPAR RNVLPSRGAS STRRARPLER PATPVPVAPS SRAARSSHIA PLLQSSSPT Highest antigen sequence indentity to the following orthologs: Mouse - 70%, Rat - 74%.

This gene encodes a member of the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcription factors. The sequence of the encoded protein contains a conserved bHLH and orange domain, but its YRPW motif has diverged from other HESR family members. It is thought to be an effector of Notch signaling and a regulator of cell fate decisions. Alternatively spliced transcript variants have been found, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEYL
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HEYL
Gene Accession No. Q9NQ87
Gene Alias bHLHb33; Class B basic helix-loop-helix protein 33; hairy and enhance of split related 3; hairy and enhancer of split-related protein 3; hairy/enhancer-of-split related with YRPW motif 3; hairy/enhancer-of-split related with YRPW motif-like; hairy/enhancer-of-split related with YRPW motif-like protein; Hairy-related transcription factor 3; hes related family bHLH transcription factor with YRPW motif-like; hesr3; hes-related family bHLH transcription factor with YRPW motif-like; HEY3; HEYL; HEY-like protein; hHeyL; hHRT3; Hrt3; HRT-3; mHRT3
Gene Symbols HEYL
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human HEYL. Recombinant protein control fragment (Product #RP-94751).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26508
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.