Learn More
HF Scientific™ Chlorine Micro Check Test Strips
Description
Includes
50 test strips are supplied in bottle labeled with instructions, color-comparison scale, and expiration date
Detect free or total chlorine concentrations as low as 0.05ppm
Test strips for field testing and quality-control procedures; provide a safe and economical alternative to reagent chlorine testing.
- Accurate results produced in less than one minute
- Easy to use—simply dip strip into sample or hold under liquid stream to saturate, remove, and match color to the color-comparison scale
Specifications
Specifications
| Format | Strip |
| For Use With (Application) | For detection of total chlorine, field testing and quality-control procedures, provide a safe and economical alternative to reagent chlorine testing |
| Includes | 50 test strips are supplied in bottle labeled with instructions, color comparison scale, and expiration date |
| Quantity | 50 Pack |
| Range | 0, 0.1, 0.2, 0.4, 0.6, 0.8, 1.2, 1.5, 2.0, 2.6, 4.0, 6.0, 10 |
| Test Time | >1 min. |
| Type | Free ChlorineTest Strip |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.