Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HF Scientific™ Chlorine Pocket Photometer

Catalog No. p-196474
Encompass
Click to view available options
Type:
Pocket Photometer (Total Cl)

Includes

Unit includes four measuring cuvets with caps, Powder Pop Dispenser with 400 doses of DPD, four AAA batteries, and instructions.

Test Free or Total Chlorine anywhere in the field for sample sizes up to 5mL

Easy-to-use microprocessor-controlled photometer provides ±2% accuracy, three-second response time, and 0.01ppm resolution.

  • Testing range: 0 to 10ppm, optical path: 25mm
  • Step-by-step menu on 16-digit alphanumeric LCD guides user through measuring process
  • Keypad protected by splash-resistant tactile membrane
  • PPD-2 DPD Powder Pop™ Dispenser provides precise, convenient push-button reagent dispensing to sample
  • Unit automatically shuts off after five minutes when not in use
  • Four “AAA” batteries provide enough power for up to 1000 tests

Order Info

Overall dimensions, 3L x 7W x 10.5 in. H (7.6 x 17.8 x 27cm). Powder Pop Dispenser also available separately—see Dispensers section.

Compliance

Meets EPA approval for measuring residual chlorine according to Standard Method 4500-Cl G. DPD Colorimetric Method.

Specifications

Accuracy ±2%
Certifications/Compliance CE, EPA 4500-ClO2D
Battery Type 4 x AAA
Display LCD
Dimensions (L x W x H) 3 x 7 x 10.5 in. (7.6 x 17.8 x 27 cm)
IP Rating IP67
Light Source LED
Type Pocket Photometer (Total Cl)
Height (English) 10.5 in.
Height (Metric) 27 cm
Includes Four measuring cuvettes with caps, Powder Pop Dispenser with 400 doses of DPD, four “AAA” batteries and instructions
Length (English) 3 in.
Length (Metric) 7.6 cm
Range 0 to 10 mg/L
Resolution 0.01 ppm
Temperature Range (Metric) 0°C to 50°C
Warranty 1 Year
Wavelength 515 nm
Width (English) 7 in.
Width (Metric) 17.8 cm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.