Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HF Scientific™ Powder Pop™ DPD Reagent for Chlorine Testing

Catalog No. 23285184
Encompass
Click to view available options
Quantity:
1000/Pk.
200/Pk.
Volume (Metric):
5 mL
10 mL
25 mL
For Use With (Application):
For testing free chlorine
For testing total chlorine

Catalog No. 23-285184

Supplier: HF Scientific™ 10303

Only null left
Add to Cart
Add to Cart

For testing free or total chlorine

  • Deliver doses of DPD reagent measured for 5, 10 or 25mL (0.2, 0.3 or 0.8 oz.) testing samples
  • Individually packaged

Specifications

Volume (Metric) 5 mL
For Use With (Application) For testing total chlorine
Quantity 200/Pk.
Type DPD Reagent
Volume (English) 0.16 oz.
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.