Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HFBI Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117598
Description
This antibody has been tested in direct-ELISA.
Contributes to surface hydrophobicity, which is important for processes such as association of hyphae in reproductive structures, dispersal of aerial spores and adhesion of pathogens to host structures.
Specifications
| HFBI | |
| Polyclonal | |
| Unconjugated | |
| hfb1; Hydrophobin I; Hydrophobin-1 | |
| Recombinant Hypocrea jecorina Hydrophobin-1 protein (23-97aa). | |
| 100 μg | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| Fungi | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction