Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HFE Antibody (SR1669), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322456100UL
Description
HFE Monoclonal antibody specifically detects HFE in Human samples. It is validated for Western BlotSpecifications
HFE | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1669 | |
Western Blot 1:500-1:2000 | |
dJ221C16.10.1, hemochromatosis, hereditary hemochromatosis protein, hereditary hemochromatosis protein HLA-H, HH, high Fe, HLAH, HLA-HHFE1, MGC103790, MHC class I-like protein HFE, MVCD7 | |
A synthesized peptide derived from human HFE (Uniprot #: Q30201) | |
100 μg | |
Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cells | |
3077 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction